Assay Details
| Gene Symbol: mtss1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044926.1 | NP_001038391.1 | 13 - 14 | 1622 | 65 | |
| XM_005169944.4 | XP_005170001.1 | 15 - 16 | 2000 | 65 | ||
| XM_005169945.4 | XP_005170002.1 | 14 - 15 | 1842 | 65 | ||
| XM_009293500.3 | XP_009291775.1 | 14 - 15 | 1983 | 65 | ||
| GenBank mRNA | BC171494.1 | - | 15 - 16 | 1802 | 65 | |
More Information
Set Membership: |
|

mtss1la
Genomic Map