Assay Details
| Gene Symbol: ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045253.1 | NP_001038718.1 | 3 - 4 | 336 | 130 | |
| GenBank mRNA | BC116616.1 | - | 3 - 4 | 336 | 130 | |
More Information
Set Membership: |
|

ngrn
Genomic Map