Assay Details
| Gene Symbol: commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077718.1 | NP_001071186.1 | 4 - 5 | 542 | 79 | |
| GenBank mRNA | BC095085.1 | - | 4 - 5 | 542 | 79 | |
| BC125866.1 | - | 4 - 5 | 542 | 79 | ||
More Information
Set Membership: |
|

commd8
Genomic Map