Assay Details
| Gene Symbol: sesn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079975.2 | NP_001073444.1 | 4 - 5 | 590 | 60 | |
| GenBank mRNA | BC134176.1 | - | 4 - 5 | 516 | 60 | |
| DQ858216.2 | - | 4 - 5 | 572 | 60 | ||
| EH536404.1 | - | 4 - 5 | 614 | 60 | ||
| EH536519.1 | - | 4 - 5 | 611 | 60 | ||
More Information
Set Membership: |
|

sesn2
Genomic Map