Assay Details
| Gene Symbol: sept8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083566.1 | NP_001077035.1 | 2 - 3 | 150 | 119 | |
| XM_005169631.3 | XP_005169688.1 | 3 - 4 | 276 | 119 | ||
| XM_021481055.1 | XP_021336730.1 | 2 - 3 | 133 | 119 | ||
| GenBank mRNA | BC095810.1 | - | 2 - 3 | 184 | 119 | |
| BC115182.1 | - | 2 - 3 | 150 | 119 | ||
More Information
Set Membership: |
|

sept8b
Genomic Map