Assay Details
| Gene Symbol: sept8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083566.1 | NP_001077035.1 | 5 - 6 | 700 | 95 | |
| XM_005169631.3 | XP_005169688.1 | 6 - 7 | 826 | 95 | ||
| XM_021481055.1 | XP_021336730.1 | 5 - 6 | 683 | 95 | ||
| GenBank mRNA | BC095810.1 | - | 5 - 6 | 734 | 95 | |
| BC115182.1 | - | 5 - 6 | 700 | 95 | ||
More Information
Set Membership: |
|

sept8b
Genomic Map