Assay Details
| Gene Symbol: sept8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083566.1 | NP_001077035.1 | 8 - 9 | 1094 | 62 | |
| XM_005169631.3 | XP_005169688.1 | 9 - 10 | 1220 | 62 | ||
| XM_021481055.1 | XP_021336730.1 | 8 - 9 | 1077 | 62 | ||
| GenBank mRNA | BC115182.1 | - | 8 - 9 | 1094 | 62 | |
More Information
Set Membership: |
|

sept8b
Genomic Map