Assay Details
| Gene Symbol: prr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110405.1 | NP_001103875.1 | 1 - 2 | 266 | 85 | |
| GenBank mRNA | BC154836.1 | - | 1 - 2 | 266 | 85 | |
More Information
Set Membership: |
|

prr16
Genomic Map