Assay Details
| Gene Symbol: lft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130960.1 | NP_571035.1 | 3 - 4 | 787 | 66 | |
| GenBank mRNA | AF132444.1 | - | 3 - 4 | 770 | 66 | |
| AF172089.1 | - | 3 - 4 | 787 | 66 | ||
More Information
Set Membership: |
|

lft1
Genomic Map