Assay Details
| Gene Symbol: jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170761.3 | NP_739567.3 | 4 - 5 | 1184 | 66 | |
| GenBank mRNA | AF401484.1 | - | 4 - 5 | 938 | 66 | |
| BC057498.1 | - | 4 - 5 | 1184 | 66 | ||
More Information
Set Membership: |
|

jmjd6
Genomic Map