Assay Details
| Gene Symbol: srpk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199650.1 | NP_955944.1 | 14 - 15 | 1672 | 69 | |
| GenBank mRNA | BC056825.1 | - | 14 - 15 | 1672 | 69 | |
More Information
Set Membership: |
|

srpk1a
Genomic Map