Assay Details
| Gene Symbol: rasd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200532.1 | NP_956826.1 | 1 - 2 | 304 | 65 | |
| GenBank mRNA | BC056272.1 | - | 1 - 2 | 304 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rasd1| Gene Symbol: rasd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200532.1 | NP_956826.1 | 1 - 2 | 304 | 65 | |
| GenBank mRNA | BC056272.1 | - | 1 - 2 | 304 | 65 | |
Set Membership: |
|