Assay Details
| Gene Symbol: cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018120.2 | NP_001018130.1 | 3 - 4 | 309 | 126 | |
| NM_200632.2 | NP_956926.1 | 3 - 4 | 309 | 126 | ||
| GenBank mRNA | AY398322.1 | - | 3 - 4 | 302 | 126 | |
| AY865566.1 | - | 2 - 3 | 174 | 126 | ||
| BC057415.1 | - | 2 - 3 | 210 | 126 | ||
| EH509759.1 | - | 3 - 4 | 289 | 126 | ||
More Information
Set Membership: |
|

cdc42
Genomic Map