Assay Details
| Gene Symbol: lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200749.2 | NP_957043.1 | 8 - 9 | 560 | 70 | |
| XM_005159779.3 | XP_005159836.1 | 9 - 10 | 1196 | 70 | ||
| XM_021468016.1 | XP_021323691.1 | 9 - 10 | 569 | 70 | ||
| GenBank mRNA | BC059556.1 | - | 7 - 8 | 489 | 70 | |
| BC159221.1 | - | 8 - 9 | 506 | 70 | ||
| EH536698.1 | - | 8 - 9 | 596 | 70 | ||
More Information
Set Membership: |
|

lypla2
Genomic Map