Assay Details
| Gene Symbol: cox4i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200803.1 | NP_957097.1 | 4 - 5 | 476 | 79 | |
| GenBank mRNA | BC059669.1 | - | 4 - 5 | 476 | 79 | |
More Information
Set Membership: |
|

cox4i2
Genomic Map