Assay Details
| Gene Symbol: relch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200844.1 | NP_957138.1 | 11 - 12 | 1672 | 98 | |
| XM_017353806.2 | XP_017209295.1 | 13 - 14 | 2796 | 98 | ||
| XM_017353807.2 | XP_017209296.1 | 13 - 14 | 2797 | 98 | ||
| XM_017353808.2 | XP_017209297.1 | 13 - 14 | 2796 | 98 | ||
| XM_017353809.2 | XP_017209298.1 | 12 - 13 | 2772 | 98 | ||
| XM_017353810.2 | XP_017209299.1 | 12 - 13 | 2773 | 98 | ||
| XM_017353813.2 | XP_017209302.1 | 11 - 12 | 2700 | 98 | ||
| GenBank mRNA | BC061956.1 | - | 11 - 12 | 1672 | 98 | |
More Information
Set Membership: |
|

relch
Genomic Map