Assay Details
| Gene Symbol: relch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200844.1 | NP_957138.1 | 18 - 19 | 2575 | 104 | |
| XM_017353806.2 | XP_017209295.1 | 20 - 21 | 3699 | 104 | ||
| XM_017353807.2 | XP_017209296.1 | 20 - 21 | 3700 | 104 | ||
| XM_017353808.2 | XP_017209297.1 | 20 - 21 | 3696 | 104 | ||
| XM_017353809.2 | XP_017209298.1 | 19 - 20 | 3675 | 104 | ||
| XM_017353810.2 | XP_017209299.1 | 19 - 20 | 3676 | 104 | ||
| XM_017353813.2 | XP_017209302.1 | 18 - 19 | 3603 | 104 | ||
| GenBank mRNA | BC061956.1 | - | 18 - 19 | 2575 | 104 | |
More Information
Set Membership: |
|

relch
Genomic Map