Assay Details
| Gene Symbol: relch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200844.1 | NP_957138.1 | 20 - 21 | 2703 | 114 | |
| XM_017353806.2 | XP_017209295.1 | 22 - 23 | 114 | |||
| XM_017353807.2 | XP_017209296.1 | 22 - 23 | 114 | |||
| XM_017353808.2 | XP_017209297.1 | 22 - 23 | 114 | |||
| XM_017353809.2 | XP_017209298.1 | 21 - 22 | 114 | |||
| XM_017353810.2 | XP_017209299.1 | 21 - 22 | 114 | |||
| XM_017353813.2 | XP_017209302.1 | 20 - 21 | 114 | |||
| GenBank mRNA | BC061956.1 | - | 20 - 21 | 2703 | 114 | |
More Information
Set Membership: |
|

relch
Genomic Map