Assay Details
| Gene Symbol: relch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200844.1 | NP_957138.1 | 24 - 25 | 3062 | 71 | |
| XM_017353806.2 | XP_017209295.1 | 26 - 27 | 4186 | 71 | ||
| XM_017353807.2 | XP_017209296.1 | 26 - 27 | 4187 | 71 | ||
| XM_017353808.2 | XP_017209297.1 | 26 - 27 | 4183 | 71 | ||
| XM_017353809.2 | XP_017209298.1 | 25 - 26 | 4162 | 71 | ||
| XM_017353810.2 | XP_017209299.1 | 25 - 26 | 4163 | 71 | ||
| XM_017353813.2 | XP_017209302.1 | 24 - 25 | 4090 | 71 | ||
| GenBank mRNA | BC061956.1 | - | 24 - 25 | 3062 | 71 | |
More Information
Set Membership: |
|

relch
Genomic Map