Assay Details
| Gene Symbol: slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200859.1 | NP_957153.1 | 8 - 9 | 974 | 79 | |
| GenBank mRNA | BC062851.1 | - | 8 - 9 | 974 | 79 | |
More Information
Set Membership: |
|

slc25a20
Genomic Map