Assay Details
| Gene Symbol: slc30a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200879.1 | NP_957173.1 | 1 - 2 | 777 | 105 | |
| GenBank mRNA | BC063939.1 | - | 1 - 2 | 777 | 105 | |
More Information
Set Membership: |
|

slc30a1a
Genomic Map