Assay Details
| Gene Symbol: rcor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205638.1 | NP_991201.1 | 8 - 9 | 949 | 67 | |
| XM_005172263.4 | XP_005172320.1 | 8 - 9 | 804 | 67 | ||
| XM_005172264.3 | XP_005172321.1 | 7 - 8 | 1041 | 67 | ||
| XM_005172265.4 | XP_005172322.1 | 7 - 8 | 658 | 67 | ||
| GenBank mRNA | BC065331.1 | - | 8 - 9 | 949 | 67 | |
More Information
Set Membership: |
|

rcor2
Genomic Map