Assay Details
| Gene Symbol: cnot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213479.1 | NP_998644.1 | 6 - 7 | 836 | 64 | |
| XM_005173877.3 | XP_005173934.1 | 6 - 7 | 954 | 64 | ||
| GenBank mRNA | BC055263.1 | - | 6 - 7 | 836 | 64 | |
| BC153627.1 | - | 6 - 7 | 848 | 64 | ||
More Information
Set Membership: |
|

cnot8
Genomic Map