Assay Details
| Gene Symbol: rpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002155.1 | NP_001002155.1 | 5 - 6 | 443 | 75 | |
| GenBank mRNA | BC071354.1 | - | 5 - 6 | 443 | 75 | |
More Information
Set Membership: |
|

rpl21
Genomic Map