Assay Details
| Gene Symbol: mob3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002191.1 | NP_001002191.1 | 1 - 2 | 439 | 75 | |
| XM_005155947.4 | XP_005156004.1 | 2 - 3 | 823 | 75 | ||
| GenBank mRNA | BC072711.1 | - | 1 - 2 | 439 | 75 | |
| BC154579.1 | - | 2 - 3 | 706 | 75 | ||
More Information
Set Membership: |
|

mob3c
Genomic Map