Assay Details
| Gene Symbol: sec61g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002499.3 | NP_001002499.2 | 3 - 4 | 368 | 79 | |
| GenBank mRNA | BC076302.1 | - | 3 - 4 | 302 | 79 | |
| BI673343.1 | - | 1 - 2 | 173 | 79 | ||
| EH544545.1 | - | 3 - 4 | 403 | 79 | ||
More Information
Set Membership: |
|

sec61g
Genomic Map