Assay Details
| Gene Symbol: zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002602.1 | NP_001002602.1 | 7 - 8 | 961 | 72 | |
| GenBank mRNA | BC075993.1 | - | 7 - 8 | 961 | 72 | |
More Information
Set Membership: |
|

zdhhc7
Genomic Map