Assay Details
| Gene Symbol: peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002616.1 | NP_001002616.1 | 5 - 6 | 916 | 64 | |
| GenBank mRNA | BC075973.1 | - | 5 - 6 | 916 | 64 | |
More Information
Set Membership: |
|

peli2
Genomic Map