Assay Details
| Gene Symbol: gna15.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003626.2 | NP_001003626.2 | 4 - 5 | 822 | 92 | |
| XM_005170450.4 | XP_005170507.1 | 4 - 5 | 975 | 92 | ||
| GenBank mRNA | BC077141.1 | - | 4 - 5 | 828 | 92 | |
More Information
Set Membership: |
|

gna15.1
Genomic Map