Assay Details
| Gene Symbol: gna15.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003626.2 | NP_001003626.2 | 7 - 8 | 1227 | 124 | |
| GenBank mRNA | BC077141.1 | - | 7 - 8 | 1233 | 124 | |
More Information
Set Membership: |
|

gna15.1
Genomic Map