Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 10 - 11 | 1522 | 98 | |
| GenBank mRNA | AY731180.1 | - | 10 - 11 | 1538 | 98 | |
| BC125892.1 | - | 10 - 11 | 1501 | 98 | ||
| CN326697.1 | - | 3 - 4 | 271 | 98 | ||
More Information
Set Membership: |
|

gsna
Genomic Map