Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 12 - 13 | 1872 | 64 | |
| XM_021468752.1 | XP_021324427.1 | 13 - 14 | 1923 | 64 | ||
| GenBank mRNA | AY731180.1 | - | 12 - 13 | 1888 | 64 | |
| BC125892.1 | - | 12 - 13 | 1851 | 64 | ||
| CN326697.1 | - | 5 - 6 | 621 | 64 | ||
| EH462972.1 | - | 4 - 5 | 664 | 64 | ||
More Information
Set Membership: |
|

gsna
Genomic Map