Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 13 - 14 | 1983 | 77 | |
| XM_021468752.1 | XP_021324427.1 | 14 - 15 | 2034 | 77 | ||
| GenBank mRNA | AY731180.1 | - | 13 - 14 | 1999 | 77 | |
| BC125892.1 | - | 13 - 14 | 1962 | 77 | ||
More Information
Set Membership: |
|

gsna
Genomic Map