Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 15 - 16 | 2127 | 63 | |
| XM_021468752.1 | XP_021324427.1 | 16 - 17 | 2178 | 63 | ||
| GenBank mRNA | AY731180.1 | - | 15 - 16 | 2143 | 63 | |
| BC125892.1 | - | 15 - 16 | 2106 | 63 | ||
| EH462972.1 | - | 1 - 2 | 408 | 63 | ||
More Information
Set Membership: |
|

gsna
Genomic Map