Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 2 - 3 | 304 | 109 | |
| XM_021468752.1 | XP_021324427.1 | 2 - 3 | 328 | 109 | ||
| GenBank mRNA | AY731180.1 | - | 2 - 3 | 317 | 109 | |
| BC115120.1 | - | 2 - 3 | 260 | 109 | ||
| BC125892.1 | - | 2 - 3 | 283 | 109 | ||
| BC134994.1 | - | 2 - 3 | 282 | 109 | ||
| EH436982.1 | - | 2 - 3 | 313 | 109 | ||
| EH457619.1 | - | 2 - 3 | 316 | 109 | ||
More Information
Set Membership: |
|

gsna
Genomic Map