Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 3 - 4 | 458 | 86 | |
| XM_021468752.1 | XP_021324427.1 | 3 - 4 | 482 | 86 | ||
| GenBank mRNA | AY731180.1 | - | 3 - 4 | 471 | 86 | |
| BC115120.1 | - | 3 - 4 | 414 | 86 | ||
| BC125892.1 | - | 3 - 4 | 437 | 86 | ||
| BC134994.1 | - | 3 - 4 | 436 | 86 | ||
| EH436982.1 | - | 3 - 4 | 467 | 86 | ||
| EH457619.1 | - | 3 - 4 | 470 | 86 | ||
More Information
Set Membership: |
|

gsna
Genomic Map