Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 4 - 5 | 622 | 110 | |
| XM_021468752.1 | XP_021324427.1 | 4 - 5 | 646 | 110 | ||
| GenBank mRNA | AY731180.1 | - | 4 - 5 | 635 | 110 | |
| BC115120.1 | - | 4 - 5 | 578 | 110 | ||
| BC125892.1 | - | 4 - 5 | 601 | 110 | ||
| BC134994.1 | - | 4 - 5 | 600 | 110 | ||
| EH436982.1 | - | 4 - 5 | 631 | 110 | ||
| EH457619.1 | - | 4 - 5 | 634 | 110 | ||
More Information
Set Membership: |
|

gsna
Genomic Map