Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 5 - 6 | 765 | 70 | |
| XM_021468752.1 | XP_021324427.1 | 5 - 6 | 789 | 70 | ||
| GenBank mRNA | AY731180.1 | - | 5 - 6 | 778 | 70 | |
| BC115120.1 | - | 5 - 6 | 721 | 70 | ||
| BC125892.1 | - | 5 - 6 | 744 | 70 | ||
| BC134994.1 | - | 5 - 6 | 743 | 70 | ||
| EH457619.1 | - | 5 - 6 | 777 | 70 | ||
More Information
Set Membership: |
|

gsna
Genomic Map