Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 6 - 7 | 866 | 105 | |
| XM_021468752.1 | XP_021324427.1 | 6 - 7 | 890 | 105 | ||
| GenBank mRNA | AY731180.1 | - | 6 - 7 | 879 | 105 | |
| BC115120.1 | - | 6 - 7 | 822 | 105 | ||
| BC125892.1 | - | 6 - 7 | 845 | 105 | ||
More Information
Set Membership: |
|

gsna
Genomic Map