Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 7 - 8 | 996 | 133 | |
| XM_021468752.1 | XP_021324427.1 | 7 - 8 | 1020 | 133 | ||
| GenBank mRNA | AY731180.1 | - | 7 - 8 | 133 | ||
| BC125892.1 | - | 7 - 8 | 975 | 133 | ||
| BC134994.1 | - | 7 - 8 | 134 | |||
More Information
Set Membership: |
|

gsna
Genomic Map