Assay Details
| Gene Symbol: gsna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012312.2 | NP_001012312.2 | 9 - 10 | 1431 | 83 | |
| XM_021468752.1 | XP_021324427.1 | 9 - 10 | 1455 | 83 | ||
| GenBank mRNA | AY731180.1 | - | 9 - 10 | 1447 | 83 | |
| BC125892.1 | - | 9 - 10 | 1410 | 83 | ||
| BC134994.1 | - | 9 - 10 | 1410 | 83 | ||
| CN326697.1 | - | 2 - 3 | 180 | 83 | ||
More Information
Set Membership: |
|

gsna
Genomic Map