Assay Details
| Gene Symbol: fn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013261.1 | NP_001013279.1 | 28 - 29 | 4540 | 83 | |
| XM_005160284.3 | XP_005160341.1 | 28 - 29 | 4572 | 83 | ||
| XM_009305554.3 | XP_009303829.1 | 29 - 30 | 4852 | 83 | ||
| GenBank mRNA | AY538257.1 | - | 28 - 29 | 4540 | 83 | |
| AY725818.1 | - | 29 - 30 | 4831 | 83 | ||
More Information
Set Membership: |
|

fn1b
Genomic Map