Assay Details
| Gene Symbol: ikbkg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014344.2 | NP_001014366.1 | 4 - 5 | 789 | 79 | |
| XM_005162070.4 | XP_005162127.1 | 4 - 5 | 803 | 79 | ||
| GenBank mRNA | AY735398.1 | - | 3 - 4 | 539 | 79 | |
| BC091820.1 | - | 4 - 5 | 787 | 79 | ||
| BC091829.1 | - | 4 - 5 | 790 | 79 | ||
| BC165252.1 | - | 3 - 4 | 570 | 79 | ||
More Information
Set Membership: |
|

ikbkg
Genomic Map