Assay Details
| Gene Symbol: hepacama | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020690.1 | NP_001018526.1 | 3 - 4 | 801 | 86 | |
| GenBank mRNA | BC095782.1 | - | 3 - 4 | 801 | 86 | |
| BC165566.1 | - | 3 - 4 | 748 | 86 | ||
More Information
Set Membership: |
|

hepacama
Genomic Map