Assay Details
| Gene Symbol: psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024441.1 | NP_001019612.1 | 7 - 8 | 589 | 88 | |
| GenBank mRNA | BC095382.1 | - | 7 - 8 | 589 | 88 | |
More Information
Set Membership: |
|

psma2
Genomic Map