Assay Details
| Gene Symbol: cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040308.1 | NP_001035398.1 | 3 - 4 | 400 | 82 | |
| XM_005165735.4 | XP_005165792.1 | 2 - 3 | 506 | 82 | ||
| GenBank mRNA | BC115279.1 | - | 3 - 4 | 400 | 82 | |
More Information
Set Membership: |
|

cdk15
Genomic Map