Assay Details
| Gene Symbol: atp2b1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044757.1 | NP_001038222.1 | 18 - 19 | 3348 | 79 | |
| XM_009300256.3 | XP_009298531.1 | 18 - 19 | 3923 | 79 | ||
| XM_021474964.1 | XP_021330639.1 | 18 - 19 | 3429 | 79 | ||
| XM_021474965.1 | XP_021330640.1 | 18 - 19 | 3429 | 79 | ||
| XM_021474966.1 | XP_021330641.1 | 17 - 18 | 3083 | 79 | ||
| GenBank mRNA | BC171622.1 | - | 17 - 18 | 3185 | 79 | |
| DQ242508.1 | - | 17 - 18 | 3307 | 79 | ||
| HM449162.1 | - | 17 - 18 | 3062 | 79 | ||
More Information
Set Membership: |
|

atp2b1a
Genomic Map