Assay Details
| Gene Symbol: znf704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044918.2 | NP_001038383.2 | 1 - 2 | 588 | 51 | |
| GenBank mRNA | BC171645.1 | - | 1 - 2 | 528 | 51 | |
| BI881870.1 | - | 1 - 2 | 309 | 51 | ||
More Information
Set Membership: |
|

znf704
Genomic Map