Assay Details
| Gene Symbol: znf704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044918.2 | NP_001038383.2 | 5 - 6 | 1264 | 63 | |
| GenBank mRNA | BC171645.1 | - | 5 - 6 | 1204 | 63 | |
More Information
Set Membership: |
|

znf704
Genomic Map