Assay Details
| Gene Symbol: mideasa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045193.1 | NP_001038658.1 | 10 - 11 | 3105 | 109 | |
| XM_005160589.4 | XP_005160646.1 | 10 - 11 | 3292 | 109 | ||
More Information
Set Membership: |
|

mideasa
Genomic Map